ContactUsLinkComponent
Fault Model
Fault Model
Catalog # 470339-416
undefined
Fault Model
Catalog # 470339-416
Supplier Number:  FAULT01
Specifications
  • Description:
    Fault model
  • Cat. no.:
    470339-416
Specifications
About this item
Demonstrate how faulting occurs in the Earth's crust.
  • Foam manipulatives
  • Demonstrate various faults
  • Access to downloadable interactive content
Set of 6 3-D manipulatives for demonstrating normal faulting, reverse (thrust) faulting and horizontal faulting. Set is provided with an access code to download interactive Teacher and Student guides, videos, geoinformation data sets, and other resource information.