Demonstration Balance Support
Demonstration balance support
Catalog # 470339-398
Supplier: United Scientific Supplies
Supplier #: DBS002
Jump to:
Product Details & Documents
Sturdy metal support for balance experiments.
- Sturdy support for meter stick demonstration balances and torque experiments
- Cast metal support measures 18.5 cm tall
Requires knife-edge clamps, slotted weights and hangers, and a meter stick (not included).
Specifications
- Description:Demonstration balance support