UniStore™ Reagent Bottles, Narrow Mouth, HDPE, 90 ml
Catalog #: 470349-792
Supplier: United Scientific Supplies
Specifications
- Capacity:90 ml
- Description:Narrow mouth reagent bottle
- Material:HDPE
Specifications
About this item
A narrow mouth HDPE bottle supplied with a lined polypropylene lid.
- Narrow mouth reagent bottles with 90 ml capacity
- Made from HDPE
- Bottles are translucent, pliable, and strong
UniStore™ narrow mouth bottle has a capacity of 90 ml and a lined polypropylene lid. Constructed of high density polyethylene, the bottle is translucent, pliable, and has excellent strength. Leakproof bottle designed to meet TSA 3-1-1 guidelines for domestic and international travel.
Ordering information: Lined polypropylene caps included.
Customers Who Bought This Also Bought