Large Laboratory Tray, Polypropylene
470339-766
- Description:Large laboratory tray, polypropylene
- Cat. no.:470339-766
A deep polypropylene tray designed for all labware needs.
- Autoclavable
- Deep 5" tray
- Tray dimensions: 20×17×5"
Large polypropylene molded laboratory tray is steam autoclavable and can be used for sterilizing and drying labware. The 5" depth makes this tray ideal for storage of labware.