Microcentrifuge Tube Rack, Polypropylene
Catalog # 470339-614
Supplier: United Scientific Supplies
Specifications
- Description:Microcentrifuge tube rack, polypropylene
- Cat. no.:470339-614
Specifications
About this item
Convenient rack goes from freezer to water bath.
- Two-tier polypropylene rack
- Autoclavable
- Use in water bath or freezers
Two-tier polypropylene racks hold up to one hundred (100) 1.5 ml microcentrifuge tubes. Racks are autoclavable and can be used in water baths or freezers. A molded-in alpha numeric index makes sample identification easy. Racks are stackable and have a 2.75×1" labeling area on two sides.
Frequently Bought Together