Order Entry
Canada
ContactUsLinkComponent
 
 
DTBP (Dimethyl 3,3′-dithiopropionimidate dihydrochloride), Pierce™
  CAPI20665
 :  
undefined
 
DTBP (Dimethyl 3,3′-dithiopropionimidate dihydrochloride), Pierce™
  CAPI20665
 :  20665
 :  

 

  • Size:
    1 g
  • Cat. no.:
    CAPI20665
  • Supplier No.:
    20665

 

 

  • Product Line:
    Pierce™
  • Labeling Method:
    Chemical Labeling
  • Crosslinker Type:
    Homobifunctional
  • Reactive Moiety:
    Imidoester
  • Spacer:
    Cleavable, Mid-Length
  • Form:
    Powder
  • Quantity:
    1 g
  • Type:
    Crosslinker
  • Solubility:
    Water

 

 

 :
 :309.28 g/mol
 :GHS07
 :Refrigerator
 :38285-78-8
 :C₈H₁₆N₂O₂S₂·2HCl
 :MFCD00043245