ContactUsLinkComponent
D-Desthiobiotin, MilliporeSigma
D-Desthiobiotin, MilliporeSigma
Catalog # CA80017-072
Supplier:  MilliporeSigma
CAS Number:  
D-Desthiobiotin, MilliporeSigma
Catalog # CA80017-072
Supplier:  MilliporeSigma
Supplier Number:  71610-3
CAS Number:  
Specifications
  • Pack type:
    Plastic ampoule
  • Description:
    D-Desthiobiotin
  • Size:
    1 g
  • Cat. no.:
    CA80017-072
  • Supplier No.:
    71610-3
Specifications
About this item
D-Desthiobiotin provides a gentle elution of StrepTag® II proteins from the biotin-binding site of StrepTactin® resins. D-Desthiobiotin is offered as a lyophilised powder for preparation of elution buffer.