Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Wax
Wax, black for dissecting trays
Wax, black for dissecting trays
Wax, black for dissecting trays
Catalog # 470332-722
Supplier:
United Scientific Supplies
CAS Number:
Expand image
Wax, black for dissecting trays
Catalog # 470332-722
Supplier:
United Scientific Supplies
Supplier Number:
DSPA05-WAX
CAS Number:
Specifications
Pack type:
Aluminum tray
Size:
2.27 kg (5 lb.)
Cat. no.:
470332-722
Specifications
About this item
Product Details
:
Product Details