ContactUsLinkComponent
CPC® MicroCNX™ Series Genderless Aseptic Fitting
CPC® MicroCNX™ Series Genderless Aseptic Fitting
Supplier:  Colder Products
Certificates
undefined
CPC® MicroCNX™ Series Genderless Aseptic Fitting
Supplier:  Colder Products
Certificates
Specifications
  • Max. operating pressure:
  • 60 psi
  • 60 psi
  • 60 psi
  • Max. temperature:
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • Min. temperature:
  • 4 °C (39 °F)
  • 4 °C (39 °F)
  • 4 °C (39 °F)
  • Sterilisation:
  • Gamma and Autoclave
  • Gamma and Autoclave
  • Gamma and Autoclave
  • Regulatory approvals:
  • USP
  • USP
  • USP