Adapter Fittings, Compression to NPT(M), Brass
Supplier: MIDWEST VALVE & FITTING INC
Certificates
About this item
Create a secure connection with compression fittings.
- Constructed of durable brass
Good chemical resistance. Rated to 400 psi maximum at 77 °F ( 25 °C). Usable from -85 to 482 °F (-65 to 250 °C).
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX31412-72
- MFLX31412-71
- MFLX31412-78
- MFLX31412-75
- MFLX31412-36
- MFLX31412-69
- MFLX31412-80
- MFLX31412-35
- Warranty:
- LTD life time
- LTD life time
- LTD life time
- LTD life time
- LTD life time
- LTD life time
- LTD life time
- LTD life time