Replacement PFA Compression Ferrule Nut and Assemblies
About this item
Create a secure connection with compression fittings.
- Constructed of chemically inert PFA
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
5 Options Available Below
Product Details & Documents
Create a secure connection with compression fittings.
- Constructed of chemically inert PFA
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Specifications for Product Family
Specifications
- Catalog No:
- MFLX31320-46
- MFLX31321-85
- MFLX31321-86
- MFLX31321-87
- MFLX31321-88
- MFLX31321-89
- MFLX31320-45
- MFLX31320-47
- MFLX31320-49
- MFLX31320-48
- Sterilisation:
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Warranty:
- 18 months
- 18 months
- 18 months
- 18 months
- 18 months
- 18 months
- 18 months
- 18 months
- 18 months
- 18 months
Specifications
Product Family Options
Product Information
- Connection TypeFlow patternFitting MaterialAvailabilityPrice
- MFLX31320-46Ferrule plug, PFA, ¼"Ferrule plug, PFA, ¼"Compression PlugPFADiscontinued-Specifications:CE certificate
More
Specifications
Cat. No.MFLX31320-46SterilisationAutoclave or ethylene oxide (ETO)Warranty18 monthsCE certificate - MFLX31321-85Compression Nut Assembly, PFA, ⅛"Compression Nut Assembly, PFA, ⅛"Compression Nut Assembly with FerruleStraightPFALog in for availabilityEach$34.29Specifications:
More
Specifications
Cat. No.MFLX31321-85SterilisationAutoclave or ethylene oxide (ETO)Warranty18 months - MFLX31321-86Compression Nut Assembly, PFA, ¼"Compression Nut Assembly, PFA, ¼"Compression Nut Assembly with FerruleStraightPFALog in for availabilityEach$34.29Specifications:
More
Specifications
Cat. No.MFLX31321-86SterilisationAutoclave or ethylene oxide (ETO)Warranty18 months - MFLX31321-87Compression Nut Assembly, PFA, ⅜"Compression Nut Assembly, PFA, ⅜"Compression Nut Assembly with FerruleStraightPFALog in for availabilityEach$41.24Specifications:
More
Specifications
Cat. No.MFLX31321-87SterilisationAutoclave or ethylene oxide (ETO)Warranty18 months - MFLX31321-88Compression Nut Assembly, PFA, ½"Compression Nut Assembly, PFA, ½"Compression Nut Assembly with FerruleStraightPFALog in for availabilityEach$44.56Specifications:
More
Specifications
Cat. No.MFLX31321-88SterilisationAutoclave or ethylene oxide (ETO)Warranty18 months - MFLX31321-89Compression Nut Assembly, PFA, ¾"Compression Nut Assembly, PFA, ¾"Compression Nut Assembly with FerruleStraightPFALog in for availabilityEach$59.09Specifications:
More
Specifications
Cat. No.MFLX31321-89SterilisationAutoclave or ethylene oxide (ETO)Warranty18 months - MFLX31320-45Ferrule plug, PFA, ⅛"Ferrule plug, PFA, ⅛"Compression PlugPFADiscontinued-Specifications:CE certificate
More
Specifications
Cat. No.MFLX31320-45SterilisationAutoclave or ethylene oxide (ETO)Warranty18 monthsCE certificate - MFLX31320-47Compression Ferrule Plug Fitting, PFA, ⅜"; 1/EachCompression Ferrule Plug Fitting, PFA, ⅜"; 1/EachCompression PlugStraightPFADiscontinued-Specifications:CE certificate
More
Specifications
Cat. No.MFLX31320-47SterilisationAutoclave or ethylene oxide (ETO)Warranty18 monthsCE certificate - MFLX31320-49Ferrule plug, PFA, ¾"Ferrule plug, PFA, ¾"Compression PlugPFADiscontinued-Specifications:CE certificate
More
Specifications
Cat. No.MFLX31320-49SterilisationAutoclave or ethylene oxide (ETO)Warranty18 monthsCE certificate - MFLX31320-48Compression Ferrule Plug Fitting, PFA, ½"; 1/EachCompression Ferrule Plug Fitting, PFA, ½"; 1/EachCompression PlugStraightPFADiscontinued-Specifications:CE certificate
More
Specifications
Cat. No.MFLX31320-48SterilisationAutoclave or ethylene oxide (ETO)Warranty18 monthsCE certificate