Nylon Fitting Manifolds, Female Threaded
Supplier: DYNAMIC MACHINING
About this item
Come in a variety of configurations—choose the number of ports and connections to suit your needs!
- Constructed of durable nylon
Constructed of nylon for economic general applications while offering good chemial compatibility.
9 Options Available Below
Product Details & Documents
Come in a variety of configurations—choose the number of ports and connections to suit your needs!
- Constructed of durable nylon
Constructed of nylon for economic general applications while offering good chemial compatibility.
Specifications for Product Family
Specifications
- Catalog No:
- MFLX31522-31
- MFLX31522-37
- MFLX31522-39
- MFLX31522-41
- MFLX31522-43
- MFLX31522-47
- MFLX31522-49
- MFLX31522-51
- MFLX31522-55
- MFLX31522-33
- MFLX31522-45
- MFLX31522-53
- MFLX31522-57
- MFLX31522-59
- Inlet NPT(F):
- 0.25"
- 0.25"
- 0.375"
- 0.5"
- 0.25"
- 0.5"
- 0.25"
- 0.375"
- 0.25"
- 0.375"
- 0.375"
- 0.5"
- 0.375"
- 0.5"
- Max. pressure:
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- Max. temperature:
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- Min. temperature:
- –54 °C (–60 °F)
- –54 °C (–60 °F)
- –54 °C (–60 °F)
- –54 °C (–60 °F)
- –54 °C (–60 °F)
- –54 °C (–60 °F)
- –54 °C (–60 °F)
- –54 °C (–60 °F)
- –54 °C (–60 °F)
- –54 °C (–60 °F)
- –54 °C (–60 °F)
- –54 °C (–60 °F)
- –54 °C (–60 °F)
- –54 °C (–60 °F)
- Mounting hole:
- 0.17"
- 0.17"
- 0.2"
- 0.2"
- 0.17"
- 0.2"
- 0.17"
- 0.2"
- 0.17"
- 0.2"
- 0.2"
- 0.2"
- 0.2"
- 0.2"
- No. of outlets:
- 2
- 4
- 4
- 4
- 6
- 6
- 8
- 8
- 10
- 2
- 6
- 8
- 10
- 10
- Outlet NPT(F):
- 0.125"
- 0.125"
- 0.25"
- 0.375"
- 0.125"
- 0.375"
- 0.125"
- 0.25"
- 0.125"
- 0.25"
- 0.25"
- 0.375"
- 0.25"
- 0.375"
- Warranty:
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
Specifications
Product Family Options
Product Information
- MaterialLengthHeightWidthAvailabilityPrice
- MFLX31522-31Nylon manifold; 2 outlets; 1/4" inlet NPT(F)Nylon manifold; 2 outlets; 1/4" inlet NPT(F)Nylon4.45 cm25.4 mm2.54 cmLog in for availabilityEach$53.20Specifications:
More
Specifications
Cat. No.MFLX31522-31Inlet NPT(F)0.25"Max. pressure200 psiMax. temperature82 °C (180 °F)Min. temperature–54 °C (–60 °F)Mounting hole0.17"No. of outlets2Outlet NPT(F)0.125"WarrantyOut of box - MFLX31522-37Nylon manifold; 4 outlets; 1/4" inlet NPT(F)Nylon manifold; 4 outlets; 1/4" inlet NPT(F)Nylon8.26 cm25.4 mm2.54 cmLog in for availabilityEach$74.80Specifications:
More
Specifications
Cat. No.MFLX31522-37Inlet NPT(F)0.25"Max. pressure200 psiMax. temperature82 °C (180 °F)Min. temperature–54 °C (–60 °F)Mounting hole0.17"No. of outlets4Outlet NPT(F)0.125"WarrantyOut of box - MFLX31522-39Nylon manifold; 4 outlets; 3/8" inlet NPT(F)Nylon manifold; 4 outlets; 3/8" inlet NPT(F)Nylon10.48 cm31.8 mm3.18 cmLog in for availabilityEach$79.70Specifications:
More
Specifications
Cat. No.MFLX31522-39Inlet NPT(F)0.375"Max. pressure200 psiMax. temperature82 °C (180 °F)Min. temperature–54 °C (–60 °F)Mounting hole0.2"No. of outlets4Outlet NPT(F)0.25"WarrantyOut of box - MFLX31522-41Nylon manifold; 4 outlets; 1/2" inlet NPT(F)Nylon manifold; 4 outlets; 1/2" inlet NPT(F)Nylon12.07 cm38.1 mm3.81 cmLog in for availabilityEach$123.00Specifications:
More
Specifications
Cat. No.MFLX31522-41Inlet NPT(F)0.5"Max. pressure200 psiMax. temperature82 °C (180 °F)Min. temperature–54 °C (–60 °F)Mounting hole0.2"No. of outlets4Outlet NPT(F)0.375"WarrantyOut of box - MFLX31522-43Nylon manifold; 6 outlets; 1/4" inlet NPT(F)Nylon manifold; 6 outlets; 1/4" inlet NPT(F)Nylon12.07 cm25.4 mm2.54 cmLog in for availabilityEach$79.70Specifications:
More
Specifications
Cat. No.MFLX31522-43Inlet NPT(F)0.25"Max. pressure200 psiMax. temperature82 °C (180 °F)Min. temperature–54 °C (–60 °F)Mounting hole0.17"No. of outlets6Outlet NPT(F)0.125"WarrantyOut of box - MFLX31522-47Nylon manifold; 6 outlets; 1/2" inlet NPT(F)Nylon manifold; 6 outlets; 1/2" inlet NPT(F)Nylon17.15 cm38.1 mm3.81 cmLog in for availabilityEach$99.00Specifications:
More
Specifications
Cat. No.MFLX31522-47Inlet NPT(F)0.5"Max. pressure200 psiMax. temperature82 °C (180 °F)Min. temperature–54 °C (–60 °F)Mounting hole0.2"No. of outlets6Outlet NPT(F)0.375"WarrantyOut of box - MFLX31522-49Nylon manifold; 8 outlets; 1/4" inlet NPT(F)Nylon manifold; 8 outlets; 1/4" inlet NPT(F)Nylon15.88 cm25.4 mm2.54 cmLog in for availabilityEach$84.30Specifications:
More
Specifications
Cat. No.MFLX31522-49Inlet NPT(F)0.25"Max. pressure200 psiMax. temperature82 °C (180 °F)Min. temperature–54 °C (–60 °F)Mounting hole0.17"No. of outlets8Outlet NPT(F)0.125"WarrantyOut of box - MFLX31522-51Nylon manifold; 8 outlets; 3/8" inlet NPT(F)Nylon manifold; 8 outlets; 3/8" inlet NPT(F)Nylon19.37 cm31.8 mm3.18 cmLog in for availabilityEach$86.70Specifications:
More
Specifications
Cat. No.MFLX31522-51Inlet NPT(F)0.375"Max. pressure200 psiMax. temperature82 °C (180 °F)Min. temperature–54 °C (–60 °F)Mounting hole0.2"No. of outlets8Outlet NPT(F)0.25"WarrantyOut of box - MFLX31522-55Nylon manifold; 10 outlets; 1/4" inlet NPT(F)Nylon manifold; 10 outlets; 1/4" inlet NPT(F)Nylon19.69 cm25.4 mm2.54 cmLog in for availabilityEach$90.60Specifications:
More
Specifications
Cat. No.MFLX31522-55Inlet NPT(F)0.25"Max. pressure200 psiMax. temperature82 °C (180 °F)Min. temperature–54 °C (–60 °F)Mounting hole0.17"No. of outlets10Outlet NPT(F)0.125"WarrantyOut of box - MFLX31522-33Nylon manifold; 2 outlets; 3/8" inlet NPT(F)Nylon manifold; 2 outlets; 3/8" inlet NPT(F)Nylon6.03 cm31.8 mm3.18 cmDiscontinued-Specifications:
More
Specifications
Cat. No.MFLX31522-33Inlet NPT(F)0.375"Max. pressure200 psiMax. temperature82 °C (180 °F)Min. temperature–54 °C (–60 °F)Mounting hole0.2"No. of outlets2Outlet NPT(F)0.25"WarrantyOut of box - MFLX31522-45Nylon manifold; 6 outlets; 3/8" inlet NPT(F)Nylon manifold; 6 outlets; 3/8" inlet NPT(F)Nylon14.93 cm31.8 mm3.18 cmDiscontinued-Specifications:
More
Specifications
Cat. No.MFLX31522-45Inlet NPT(F)0.375"Max. pressure200 psiMax. temperature82 °C (180 °F)Min. temperature–54 °C (–60 °F)Mounting hole0.2"No. of outlets6Outlet NPT(F)0.25"WarrantyOut of box - MFLX31522-53Nylon manifold; 8 outlets; 1/2" inlet NPT(F)Nylon manifold; 8 outlets; 1/2" inlet NPT(F)Nylon22.23 cm38.1 mm3.81 cmDiscontinued-Specifications:
More
Specifications
Cat. No.MFLX31522-53Inlet NPT(F)0.5"Max. pressure200 psiMax. temperature82 °C (180 °F)Min. temperature–54 °C (–60 °F)Mounting hole0.2"No. of outlets8Outlet NPT(F)0.375"WarrantyOut of box - MFLX31522-57Nylon manifold; 10 outlets; 3/8" inlet NPT(F)Nylon manifold; 10 outlets; 3/8" inlet NPT(F)Nylon23.81 cm31.8 mm3.18 cmDiscontinued-Specifications:
More
Specifications
Cat. No.MFLX31522-57Inlet NPT(F)0.375"Max. pressure200 psiMax. temperature82 °C (180 °F)Min. temperature–54 °C (–60 °F)Mounting hole0.2"No. of outlets10Outlet NPT(F)0.25"WarrantyOut of box - MFLX31522-59Nylon manifold; 10 outlets; 1/2" inlet NPT(F)Nylon manifold; 10 outlets; 1/2" inlet NPT(F)Nylon27.31 cm38.1 mm3.81 cmDiscontinued-Specifications:
More
Specifications
Cat. No.MFLX31522-59Inlet NPT(F)0.5"Max. pressure200 psiMax. temperature82 °C (180 °F)Min. temperature–54 °C (–60 °F)Mounting hole0.2"No. of outlets10Outlet NPT(F)0.375"WarrantyOut of box