Corning® Lambda™ EliteMax Benchtop Workstation, Corning
The New Corning® LAMBDA™ EliteMax Semi-Automated Benchtop Pipettor is the optimal solution for purification, isolation, and extraction for sample prep and compound management workflows.
- Blowout feature helps to dispense all excess liquid that remains in the tip after dispensing
- Speed can be changed depending on the type of liquid or compound being transferred
- The large touch screen controller allows the user to easily control the device
- There is also an option for the instrument to be controlled manually on the touch screen controller
- You can create multiple user accounts for storing preferences, protocols, and methods
The Corning® LAMBDA™ EliteMax Semi-Automated Benchtop Pipettor is a high throughout automation platform with a small footprint for:
- Plate filling from reagents reservoirs
- Serial dilutions, plate-to-plate transfers, tube-to-tube transfers, and plate replication
- Well-to-well transfers (cherry picking): Single-channel, 12-channel, 24-channel, and custom-channel heads are available
- Accurately and precisely transferring liquids and compounds between microplates
- Transferring by row or by column
Certifications: Calibration certificate.
Packaging: Palletized wooden crate.
Documents
Corning® Lambda™ EliteMax semi-automated benchtop pipettor Corning® Lambda™ EliteMax Semi-automated Benchtop Pipettor Corning® Lambda™ EliteMax Semi-automated Benchtop Pipettor, Cherry Picking Protocol Corning® Lambda™ EliteMax Semi-automated Benchtop Pipettor, Linking Protocol Corning® Lambda™ EliteMax Semi-automated Benchtop Pipettor, Plate Definition
Videos
Manual Corning® EliteMax Service Manual Corning® Lambda™ EliteMax Semi-automated Benchtop Pipettor Quick Start Guide Corning® Lambda™ EliteMax Semi-automated Benchtop Pipettor
Manual
- Description:Corning® Lambda™ EliteMax semi-automated benchtop pipettor
- Cat. No.:76531-252
- Weight of system:44 lbs. (20 kg)
- No. of channels:1 / 12 / 24
- Model:Corning® Lambda™ EliteMax



