ContactUsLinkComponent
AssureMask® Precision Care™ Procedure Tie-On Face Masks
AssureMask® Precision Care™ Procedure Tie-On Face Masks
Catalog # CA76544-808
Supplier:  Medicom
AssureMask® Precision Care™ Procedure Tie-On Face Masks
Catalog # CA76544-808
Supplier:  Medicom
Supplier Number:  206914
Specifications
  • Sterility:
    Non sterile
  • Color:
    White
  • Description:
    AssureMask® Precision Care™ tie-on face masks, ASTM Level 1
  • Cat. no.:
    CA76544-808
Specifications
About this item
Fog-free protection with a personalized fit.
  • Non-woven spunbond outer layer provides maximum protection against fluid penetration
  • Pleated design prevents liquid buildup and cross-contamination
  • High-quality material ensures optimal filtration and breathability
  • Anti-fog nose strip based on SureView™ technology ensures an unobstructed view of the surgical field
  • Latex-free
SureView™ technology ensures an unobstructed view of the operative field.​ High-quality materials and a soft inner lining made for sensitive skin enhance breathability and comfort. Soft, strong, ultrasonically sealed tabs attached to the outside of the mask prevent pulling and chafing and allow for easy removal with less risk of self-contamination. Individually folded ties under each mask for easy dispensing.
Procedure masks are intended to be worn by personnel during medical procedures to protect both the patient and the operating personnel from transfer of microorganisms, body fluids, and particulate material transfer.
Packaging: Masks are packaged 50/box, 6 boxes/case.