Specifications
- Description:FIT mailers
- Cat. no.:76543-130
Specifications
About this item
The Rapid Response™ fecal immunochemical test (FIT) mailer is a paper kit designed to be given to patients for at-home collection of spontaneously passed stool specimens (SPS). This easy collection method increases patient compliance and ensures an appropriate sample.
- Enables easy at-home collection
- Ensures appropriate sample collection
Use in conjunction with the Rapid Response™ FIT test (VWR Cat. No. 76473-472) and the Rapid Response™ IFOBT external controls (VWR Cat. No. 76473-474).