Rapid Response™ Multi-Drug Flat Test Cup, BTNX
Supplier: BTNX INC.
Certificates
About this item
The Rapid Response™ multi-drug flat cup is an easy-to-use, one-step solution for drug screening that can simultaneously detect multiple drugs and metabolites in urine samples at specified cut-off levels.
- All in one step
- No leakage
- No contact with urine (no mess)
- Results in just 5 minutes
Interpretation of test results are: positive (one line), negative (two lines), invalid (no lines or no control line), or the comparison of reaction pad color to the corresponding color blocks for the adulteration strips.
Delivery: 25 Individually packaged test cups and the product insert.
Packaging: 1 cup per pouch.
Caution: For forensic use only.
Frequently Bought Together