J&W CP-Sil 13 CB Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Delivers guaranteed analysis of halocarbons according to EPA methods 601, 602, and 624.
- 14% phenyl/86% dimethylpolysiloxane
- Mid-polarity phase
- Specially developed for the analysis of medium-polarity compounds such as halocarbons
- Well suited for EPA Methods 601, 602, and 624
- Ideal for confirmation analysis using ECD
- Bonded, crosslinked, and solvent rinsable
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
- Similar phases: Rtx-20
Agilent J&W CP-Sil 13 CB is a 14%-phenyl/86% dimethylpolysiloxane for the analysis of halocarbons with EPA methods 601, 602, and 624, as well as other medium polarity compounds. The column offers unique sensitivity and productivity and enables separations at high flow rates, with minimum bleed when using Electronic Capture Detector (ECD), due to the absence of cyano groups.
Specifications
- Catalog No:
- CAAGCP7813
- CAAGCP7926
- Description:
- GC column, 7 inch cage
- GC column, 7 inch cage
- Polarity:
- Mid-polarity stationary phase
- Mid-polarity stationary phase