ContactUsLinkComponent
Masterflex® Reducer Fittings, Male NPT Threaded, Straight, Avantor®
Masterflex® Reducer Fittings, Male NPT Threaded, Straight, Avantor®
Supplier:  VWR International
Certificates
undefined
Masterflex® Reducer Fittings, Male NPT Threaded, Straight, Avantor®
Supplier:  VWR International
Certificates
Specifications
  • Model:
  • 30623-38
  • 30623-32
  • 30623-46
  • 30623-42
  • 30623-48
  • 40627-35
  • 40627-43
  • 40623-47
  • 40623-39
  • 40623-55
  • 30623-50
  • 40623-43
  • 40623-51
  • 30623-40
  • 30623-54
  • 40623-35
  • 30623-52
  • 30627-54
  • 31701-89
  • 31701-88
  • 30623-36
  • 30623-34
  • 40627-55
  • Sterilisation:
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) or gamma
  • Autoclave or ethylene oxide (ETO)
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Warranty:
  • 1 year
  • 1 year