ContactUsLinkComponent
Masterflex® Correlated Variable-Area Flowmeters, High-Res Valve, High-Flow, 150-mm, Avantor®
Masterflex® Correlated Variable-Area Flowmeters, High-Res Valve, High-Flow, 150-mm, Avantor®
Certificates
undefined
Masterflex® Correlated Variable-Area Flowmeters, High-Res Valve, High-Flow, 150-mm, Avantor®
Specifications
  • Model:
  • 03227-36
  • 03227-87
  • 03295-32
  • 03295-36
  • 03229-35
  • 03295-34
  • 03227-32
  • 03227-34
  • 03229-33
  • 03229-37
  • 03217-78
  • Accuracy:
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • Float material:
  • Carboloy
  • Glass
  • Carboloy
  • 316 SS
  • 316 SS
  • Glass
  • 316 SS
  • Glass
  • Carboloy
  • Sapphire
  • Flow Tube material:
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Max. operating temperature:
  • 121 °C (250 °F)
  • 65 °C (150 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 65 °C (150 °F)
  • O-Ring material:
  • Buna-N
  • PTFE
  • Buna-N
  • Buna-N
  • Viton®
  • Buna-N
  • Buna-N
  • Buna-N
  • Viton®
  • Viton®
  • PTFE
  • Repeatability:
  • ±0.25%
  • ±0.25% full-scale
  • ±0.25%
  • ±0.25%
  • ±0.25%
  • ±0.25%
  • ±0.25%
  • ±0.25%
  • ±0.25%
  • ±0.25%
  • ±0.25%
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year