ContactUsLinkComponent
Blue-White® Direct-Read Variable-Area Flowmeters for Water, Polysulfone Housing
Blue-White® Direct-Read Variable-Area Flowmeters for Water, Polysulfone Housing
Supplier:  VWR International
Certificates
undefined
Blue-White® Direct-Read Variable-Area Flowmeters for Water, Polysulfone Housing
Supplier:  VWR International
Specifications
  • Model:
  • 32472-00
  • 32470-01
  • 32472-02
  • 32473-01
  • 32470-03
  • 32473-02
  • 32470-02
  • 32473-04
  • 32472-03
  • 32472-01
  • 32470-00
  • 32470-04
  • Accuracy:
  • ±3% full-scale
  • ±4% full-scale
  • ±3% full-scale
  • ±3% full-scale
  • ±4% full-scale
  • ±3% full-scale
  • ±4% full-scale
  • ±3% full-scale
  • ±3% full-scale
  • ±3% full-scale
  • ±4% full-scale
  • ±4% full-scale
  • Float material:
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • PVC
  • 316 SS
  • Max. operating temperature:
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 65 °C (150 °F)
  • 100 °C (212 °F)
  • 65 °C (150 °F)
  • 100 °C (212 °F)
  • 65 °C (150 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year