Masterflex® Ismatec® Peristaltic Pump Footswitches, Avantor®
Simplify your dispensing.
- Select the footswitch with connectio suitable for your Ismatec® pump
Footswitches provide convenient, hands-free operation of your pump drives from up to six feet away. Simply press once to start pump, press again to stop.
- Environmentally Preferable:
- Model:
- 78003-07
- 07610-25
- 78016-91
- 78016-95
- 78016-93
- Input:
- DB-15
- 3-pin circular connector
- DB-15
- 8-Pin DIN
- DB-9
- Series:
- Ismatec BVP and MCP Drives
- Ismatec Digital Drives
- Ismatec Reglo Analog Drives
- Ismatec Ecoline Drives
- Ismatec Reglo Digital Drives
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Frequently Bought Together