ContactUsLinkComponent
Masterflex® L/S® MiniFlex® OEM Pump Heads, Avantor®
Masterflex® L/S® MiniFlex® OEM Pump Heads, Avantor®
Certificates
undefined
Masterflex® L/S® MiniFlex® OEM Pump Heads, Avantor®
Specifications
  • Model:
  • 77220-00
  • 77220-10
  • Gear ratio:
  • 1:1
  • 1:1
  • Max. flow rate per channel:
  • 315 ml/min
  • 162 ml/min
  • Min. flow rate per channel:
  • 0.004 ml/min
  • 0.004 ml/min
  • No. of channels:
  • 1
  • 2
  • Occlusion:
  • Fixed
  • Fixed
  • Open-head sensor:
  • Yes
  • Yes
  • Optimum RPM range:
  • 0.1 - 300 rpm
  • 0.1 - 300 rpm
  • Pump series:
  • L/S
  • L/S
  • Tubing retention:
  • Automatic
  • Automatic
  • Tubing sizes accepted:
  • L/S® 13, L/S 14, L/S 16, L/S 25
  • L/S® 13, L/S 14, L/S 16
  • Volume per revolution with L/S 13 tubing:
  • 0.04 ml
  • 0.04 ml
  • Volume per revolution with L/S 14 tubing:
  • 0.14 ml
  • 0.14 ml
  • Volume per revolution with L/S 16 tubing:
  • 0.5 ml
  • 0.5 ml
  • Volume per revolution with L/S 25 tubing:
  • 1.1 ml
  • Warranty:
  • 2 years
  • 2 years