ContactUsLinkComponent
Masterflex® Open-Head Sensor Retrofit System, Avantor®
Masterflex® Open-Head Sensor Retrofit System Controller
  MFLX77097-00
CE Compliant
ETL listed
ISO9001
undefined
Masterflex® Open-Head Sensor Retrofit System Controller
  MFLX77097-00
 :  77097-00
CE Compliant
ETL listed
ISO9001
  • Description:
    Open-Head Sensor Retrofit System Controller
  • Controller IP rating:
    IP 66
  • Drive requirements:
    Compatible with all I/P drives and L/S drives that accept the Easy-Load II pump head
  • Type of pump heads accepted:
    Use only with I/P Easy-Load pump head 77602-30 and L/S Easy-Load II pump heads 77200-30 and 77200-32
  • Warranty:
    2 years
  • Cat. no.:
    MFLX77097-00
  • No. of Pump Heads Accepted:
    2
Instantly shuts down the drive if the pump head is opened during operation.
  • Use in conjunction with Masterflex® L/S® and I/P® open-head sensor equipped pump heads
  • Compatible with Masterflex® L/S® and I/P® pumps without integrated open-head sensor
  • Controller accepts input signals from up to two pump heads
  • Protect technicians and operators tasked with tubing changes and routine maintenance in busy production environments
  • Controller is NEMA 4X / IP66 rated for washdown
The retrofit system is compatible with all I/P® pumps and with L/S® pumps that accept the Easy-Load® II pump head.
Use only with I/P® Easy-Load® pump head 77602-30 and with L/S® Easy-Load® II pump heads 77200-30 and 77200-32.
  • Description:
    Open-Head Sensor Retrofit System Controller
  • Environmentally Preferable:
  • Controller IP rating:
    IP 66
  • Drive requirements:
    Compatible with all I/P drives and L/S drives that accept the Easy-Load II pump head
  • Type of pump heads accepted:
    Use only with I/P Easy-Load pump head 77602-30 and L/S Easy-Load II pump heads 77200-30 and 77200-32
  • Warranty:
    2 years
  • Cat. no.:
    MFLX77097-00
  • No. of Pump Heads Accepted:
    2