ContactUsLinkComponent
Masterflex® L/S® Tang Adapter Kits, Avantor®
Masterflex® L/S® Tang Adapter Kit, 2 Large and 2 Small Tang Adapters
Catalog # MFLX07519-01
undefined
Masterflex® L/S® Tang Adapter Kit, 2 Large and 2 Small Tang Adapters
Catalog # MFLX07519-01
Supplier Number:  7519-01
Specifications
  • Pump series:
    L/S
  • Description:
    L/S Tang Adapter Kit, 2 large and 2 small per kit
  • Cat. no.:
    MFLX07519-01
Specifications
About this item
Extend the life of your Masterflex pump head with these replacement parts.
  • Suitable for use with PTFE Diaphragm pump head and Multichannel Cartridge pump heads
Replacement parts to keep your pump head operating even longer. Tang adapter kits provides a tight tang shaft connection and reduces noise. Use large adapter for 75W (1/10 hp) drives and small adapter for 37W (1/20 hp drives).
  • Pump series:
    L/S
  • Description:
    L/S Tang Adapter Kit, 2 large and 2 small per kit
  • Environmentally Preferable:
  • Cat. no.:
    MFLX07519-01