Orders LinkContactUsLinkComponent
Masterflex® L/S® Multichannel Pump Head Cartridges, Avantor®
Masterflex® L/S® Multichannel Pump Head Cartridges, Avantor®
Masterflex® L/S® Multichannel Pump Head Cartridges, Avantor®
Masterflex® L/S® Multichannel Pump Head Cartridges, Avantor®
Use with L/S® multichannel cartridge pump heads for synchronous, multichannel pumping.
  • Only Silicone and C-Flex® tubing formulations are recommended for size L/S® 17 tubing when used in L/S® cartridge pump heads.
Cartridges feature accurate flow rates and adjustable tubing retainers to hold tubing in place — for large cartridges, set tubing retainers to your tube size and you are ready to pump. On small cartridges, optimum retention is preset — simply load tubing and start pumping.
Cartridges are constructed from polycarbonate and glass-filled nylon. Mix small and large cartridges on pump heads that accept both sizes; two small cartridges occupy the same space as one large cartridge.
  • Model:
  • 07519-70
  • 07519-75
  • 07519-80
  • 07519-85
  • No. of channels:
  • 1
  • 1
  • 1
  • 1
  • Min. flow rate per channel:
  • 0.004 ml/min
  • 0.0006 ml/min
  • 0.0015 ml/min
  • 1E-05 ml/min
  • Max. flow rate per channel:
  • 1700 ml/min
  • 350 ml/min
  • 130 ml/min
  • 85 ml/min
  • Open-head sensor:
  • No
  • No
  • No
  • No
  • Pump series:
  • L/S
  • L/S
  • L/S
  • L/S
  • Tubing sizes accepted:
  • L/S® 14, L/S 16, L/S 25, L/S 17
  • L/S® 14, L/S 16, L/S 25, L/S 17
  • Microbore two-stop tubing sets, L/S 13, L/S 14
  • Microbore two-stop tubing sets
  • Occlusion:
  • Adjustable Tangential
  • Adjustable Offset Tangential
  • Adjustable Tangential
  • Adjustable Offset Tangential
  • Optimum RPM range:
  • 0.02 - 600 rpm
  • 0.02 - 600 rpm
  • 0.02 - 600 rpm
  • 0.02 - 600 rpm
  • Tubing retention:
  • Manual
  • Manual
  • Stopped Tubing or Manual
  • Stopped Tubing
  • Volume per revolution with L/S 13 tubing:
  • 0.06 ml
  • Volume per revolution with L/S 14 tubing:
  • 0.21 ml
  • 0.21 ml
  • 0.21 ml
  • Volume per revolution with L/S 16 tubing:
  • 0.6 ml
  • 0.5 ml
  • Volume per revolution with L/S 17 tubing:
  • 2.3 ml
  • 1.4 ml
  • Volume per revolution with L/S 25 tubing:
  • 1.4 ml
  • 0.96 ml
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Product Family Options
  • Description
    Maximum Flow Rate
    Height
    Length
    Width
    Housing material
  • L/S large cartridges for 07519-05 and -06 pump heads.
    1700 ml/min
    95 mm (3.75")
    125 mm (4.3125")
    19 mm (0.75")
    PC and glass-filled nylon
    Each
    Each - $223.00
    $223.00
    Each
       $223.00
     : 7519-70
    Specifications
    Cat. No.
    MFLX07519-70
    Model
    07519-70
    No. of channels
    1
    Min. flow rate per channel
    0.004 ml/min
    Max. flow rate per channel
    1700 ml/min
    Open-head sensor
    No
    Pump series
    L/S
    Tubing sizes accepted
    L/S® 14, L/S 16, L/S 25, L/S 17
    Occlusion
    Adjustable Tangential
    Optimum RPM range
    0.02 - 600 rpm
    Tubing retention
    Manual
    Volume per revolution with L/S 13 tubing
    Volume per revolution with L/S 14 tubing
    0.21 ml
    Volume per revolution with L/S 16 tubing
    0.6 ml
    Volume per revolution with L/S 17 tubing
    2.3 ml
    Volume per revolution with L/S 25 tubing
    1.4 ml
    Warranty
    1 year
  • L/S large cartridges for 07519-10 and -15 pump heads.
    350 ml/min
    95 mm (3.75")
    125 mm (4.3125")
    19 mm (0.75")
    PC and glass-filled nylon
    Each
    Each - $223.00
    $223.00
    Each
       $223.00
     : 7519-75
    Specifications
    Cat. No.
    MFLX07519-75
    Model
    07519-75
    No. of channels
    1
    Min. flow rate per channel
    0.0006 ml/min
    Max. flow rate per channel
    350 ml/min
    Open-head sensor
    No
    Pump series
    L/S
    Tubing sizes accepted
    L/S® 14, L/S 16, L/S 25, L/S 17
    Occlusion
    Adjustable Offset Tangential
    Optimum RPM range
    0.02 - 600 rpm
    Tubing retention
    Manual
    Volume per revolution with L/S 13 tubing
    Volume per revolution with L/S 14 tubing
    0.21 ml
    Volume per revolution with L/S 16 tubing
    0.5 ml
    Volume per revolution with L/S 17 tubing
    1.4 ml
    Volume per revolution with L/S 25 tubing
    0.96 ml
    Warranty
    1 year
  • L/S small cartridges for 07519-05 and -06 pump heads.
    130 ml/min
    95 mm (3.75")
    125 mm (4.3125")
    10 mm (0.375")
    PC and glass-filled nylon
    Each
    Each - $223.00
    $223.00
    Each
       $223.00
     : 7519-80
    Specifications
    Cat. No.
    MFLX07519-80
    Model
    07519-80
    No. of channels
    1
    Min. flow rate per channel
    0.0015 ml/min
    Max. flow rate per channel
    130 ml/min
    Open-head sensor
    No
    Pump series
    L/S
    Tubing sizes accepted
    Microbore two-stop tubing sets, L/S 13, L/S 14
    Occlusion
    Adjustable Tangential
    Optimum RPM range
    0.02 - 600 rpm
    Tubing retention
    Stopped Tubing or Manual
    Volume per revolution with L/S 13 tubing
    0.06 ml
    Volume per revolution with L/S 14 tubing
    0.21 ml
    Volume per revolution with L/S 16 tubing
    Volume per revolution with L/S 17 tubing
    Volume per revolution with L/S 25 tubing
    Warranty
    1 year
  • L/S small cartridges for 07519-20 and -25 pump heads.
    85 ml/min
    95 mm (3.75")
    125 mm (4.3125")
    10 mm (0.375")
    PC and glass-filled nylon
    Each
    Each - $223.00
    $223.00
    Each
       $223.00
     : 7519-85
    Specifications
    Cat. No.
    MFLX07519-85
    Model
    07519-85
    No. of channels
    1
    Min. flow rate per channel
    1E-05 ml/min
    Max. flow rate per channel
    85 ml/min
    Open-head sensor
    No
    Pump series
    L/S
    Tubing sizes accepted
    Microbore two-stop tubing sets
    Occlusion
    Adjustable Offset Tangential
    Optimum RPM range
    0.02 - 600 rpm
    Tubing retention
    Stopped Tubing
    Volume per revolution with L/S 13 tubing
    Volume per revolution with L/S 14 tubing
    Volume per revolution with L/S 16 tubing
    Volume per revolution with L/S 17 tubing
    Volume per revolution with L/S 25 tubing
    Warranty
    1 year