ContactUsLinkComponent
Masterflex® Mechanically Coupled High-Head Centrifugal Pumps, Avantor®
undefined
Masterflex® Mechanically Coupled High-Head Centrifugal Pumps, Avantor®
  • Model:
  • 70725-37
  • 70725-13
  • 70725-26
  • 70725-31
  • 70725-32
  • 70725-36
  • 70725-11
  • 70725-20
  • 70725-22
  • Duty cycle continuous:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Inlet connection:
  • 2.5" flange
  • 1.5 NPT(F)
  • 2" flange
  • 2" flange
  • 2" flange
  • 2.5" flange
  • 1.5 NPT(F)
  • 2" flange
  • 2" flange
  • Max. pressure:
  • 8.6 bar (125 psi)
  • 8.6 bar (125 psi)
  • 15.8 bar (230 psi)
  • 15.8 bar (230 psi)
  • 8.6 bar (125 psi)
  • 8.6 bar (230 psi)
  • 8.6 bar (125 psi)
  • 15.9 bar (230 psi)
  • 8.6 bar (125 psi)
  • Max. temperature:
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • Max. viscosity:
  • 352 cP
  • 352 cP
  • 352 cP
  • 352 cP
  • 352 cP
  • 352 cP
  • 352 cP
  • 352 cP
  • 352 cP
  • Motor type:
  • TEFC
  • TEFC
  • TEFC
  • TEFC
  • TEFC
  • TEFC
  • TEFC
  • TEFC
  • TEFC
  • Motor hp:
  • 7.5
  • 1-¹/₂
  • 5
  • 7.5
  • 10
  • 5
  • 1-¹/₂
  • 3
  • 3
  • Outlet connection:
  • 1.5" flange
  • 1 NPT(F)
  • 1.25" flange
  • 1.25" flange
  • 1.25" flange
  • 1.5" flange
  • 1 NPT(F)
  • 1.25" flange
  • 1.25" flange
  • Wetted materials:
  • 304 SS pump head and impeller; SS shaft; Type 21 mechanical seals (carbon vs. ceramic/Viton®); and Viton O-rings
  • 304 SS pump head and impeller; SS shaft; Type 21 mechanical seals (carbon vs. ceramic/Viton®); and Viton O-rings
  • 304 SS pump head and impeller; SS shaft; Type 21 mechanical seals (carbon vs. ceramic/Viton®); and Viton O-rings
  • 304 SS pump head and impeller; SS shaft; Type 21 mechanical seals (carbon vs. ceramic/Viton®); and Viton O-rings
  • 304 SS pump head and impeller; SS shaft; Type 21 mechanical seals (carbon vs. ceramic/Viton®); and Viton O-rings
  • 304 SS pump head and impeller; SS shaft; Type 21 mechanical seals (carbon vs. ceramic/Viton®); and Viton O-rings
  • 304 SS pump head and impeller; SS shaft; Type 21 mechanical seals (carbon vs. ceramic/Viton®); and Viton O-rings
  • 304 SS pump head and impeller; SS shaft; Type 21 mechanical seals (carbon vs. ceramic/Viton®); and Viton O-rings
  • 304 SS pump head and impeller; SS shaft; Type 21 mechanical seals (carbon vs. ceramic/Viton®); and Viton O-rings
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Frequently Bought Together