Order Entry
Canada
ContactUsLinkComponent
Whatman™ Mixed Cellulose Ester Membranes, Sterile, 0.6 µm, Whatman Products (Cytiva)
Whatman™ Mixed Cellulose Ester Membranes, Sterile, 0.6 µm, Whatman Products (Cytiva)
Supplier:  Cytiva
Certificates
undefined
Whatman™ Mixed Cellulose Ester Membranes, Sterile, 0.6 µm, Whatman Products (Cytiva)
Supplier:  Cytiva

Specifications

  • Catalog No:
  • CA11012-910
  • Max. operating temperature:
  • 130°C
  • Application:
  • The membrane is particularly effective in applications requiring higher flow rates and larger volume filtration including clarification or sterilization of aqueous solutions, particulate analysis and removal, air monitoring and microbial analysis. Other applications include:
    Clarification or sterilization of aqueous solutions
    Cytology
    Air monitoring
    HPLC samples (aqueous)
    Virus concentration
    Particulate analysis
    Biological assays
    Food microbiology including enumeration of E. coli in foods
    Bacteriological studies
    Particle counting from liquids and aerosols
    Yeasts and molds.
  • Autoclavable:
  • Yes
  • Burst strength:
  • >0.68 bar
  • Porosity:
  • 74% min. to 77% max.
  • Protein:
  • Medium Protein Binding
  • Solvent:
  • Medium Solvent Resistance

Specifications

Related Information

Frequently Bought Together