DR1300 FL Portable Fluorometer, Hach
Supplier: Hach
Certificates
About this item
Protect your RO membrane investment with ultra-low range chlorine and sulfite analysis.
- Protect Your assets
- Monitor and optimize your dechlorination process
- Depend on direct measurements
- New fluorescence technology
The DR1300 FL features new fluorescence technology only from Hach. Think about lighting bugs in a night sky. That’s what happens when you activate the glow with 10 easy drops of our specially formulated reagent. Then, you’ll follow the fluorescence to process improvements by understanding how much chlorine is in your system.
Handheld Fluorometer with innovative fluorescence technology for ultra low range free and total chlorine and sulfite testing: Free Chlorine: 2 to 100 µg/L Cl₂; Total Chlorine: 3 to 100 µg/L Cl₂; Sulfite: 6 to 500 µg/L SO₃.
The DR1300 FL is suitable for indoor and outdoor use.
Delivery: DR1300 FL fluorometer includes 4×AA batteries, sample vial adapter, sample vial adapter cover, and user manual.
Related Information
DR1300 FL_Fluorescence-INFORMATIVE How to Test for Chlorine and Sulfite in Water with Hach’s DR1300 FL
Videos
Hach’s DR1300 FL Bisulfite Dechlorination Control Hach’s DR1300 FL GAC Dechlorination Control
Application Notes
Manual Chlorine, Free, Ultra Low Range, Fluorescence Method Manual Chlorine, Total, Ultra Low Range, Fluorescence Method Manual DR1300 FL Portable Fluorometer Manual Sulfite, Ultra Low Range, Fluorescence Method
Manual
Online Membrane Monitoring and Protection from Hach
Additional Product Details
Customers Who Bought This Also Bought