Order Entry
Canada
ContactUsLinkComponent
RNA-Solv® RNA Isolation, Reagent, Omega bio-tek
RNA-Solv® RNA Isolation, Reagent, Omega bio-tek
Catalog # CA76509-254
Supplier:  Omega Bio-Tek
CAS Number:  
undefined
RNA-Solv® RNA Isolation, Reagent, Omega bio-tek
Catalog # CA76509-254
Supplier:  Omega Bio-Tek
Supplier Number:  R6830-02
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Organic extraction
  • Prep size:
    Scalable
  • Refine target molecule:
    Total RNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Description:
    E.Z.N.A.® RNA-Solv® reagent
  • Size:
    200 ml
  • Sample size:
    1×10⁷ cells or 100 mg tissue
  • Yield:
    10 - 200 µg
  • Cat. no.:
    CA76509-254

Specifications

About this item

RNA-Solv® Reagent is a one reagent system for the isolation of total RNA from cells and tissues. The reagent, a single-phase solution consisting of phenol and guanidine isothiocyanate, is a modification of the single-step RNA isolation method developed by Chomczynski and Sacchi. The sample is homogenised and lysed in RNA-Solv® Reagent, which maintains the integrity of the RNA while disrupting and denaturing endogenous RNases and other cellular components. Extraction of the lysate with chloroform further denatures proteins and separates the mixture into an organic and an aqueous phase. RNA remains exclusively in the aqueous phase, and is subsequently recovered by isopropanol.

This method is suitable for a wide range of starting material: Up to 1 g of tissue or 1×10⁸ cells of human, animal, plant, or bacterial origin. The simplicity of the RNA-Solv® Reagent method allows simultaneous processing of a large number of samples. The entire procedure can be completed in one hour. Total RNA prepared can be used for Northern blot analysis, dot blot hybridization, poly(A)+ selection, in vitro translation, RNase protection assay, and molecular cloning. For use in amplification by thermal cycling, treatment of the isolated RNA with RNase-free DNase I is recommended when the two amplimers lie within a single exon.