Orders LinkContactUsLinkComponent
Styrofoam Head Models
Styrofoam Head Models
Styrofoam Head Models
Certificates
Styrofoam Head Models
Styrofoam Head Models
3 Options Available Below
Product Details & Documents
Styrofoam head models for multiple uses.
  • Available in male, female, and faceless versions
  • Smooth surface paints easily
  • Dense and durable
These smooth foam head forms can be painted or finished, allowing students to make or display their own models. Use them as a base to create brain maps, or sculpt masks. The foam is denser than most styrofoam, so it can also be trimmed or cut to create internal anatomical models. They may also be used for basic modeling projects in multiple disciplines.
Product Family Options
Product Information
  • Size
    Availability
    Price
  • Female
    6.3×6.8×10"
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $23.60
    $23.60
    Log in for availability
    Case of 6
    Case of 6 - $134.70
    $134.70
    Each
       $23.60
    Case of 6
       $134.70
    Supplier Number: RS260/6
  • Male
    6.9×7.1×10.1"
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $23.60
    $23.60
    Log in for availability
    Case of 4
    Case of 4 - $89.90
    $89.90
    Each
       $23.60
    Case of 4
       $89.90
    Supplier Number: RS261/4
  • 470337-668
    Faceless
    Faceless
    5.8×6.8×9.8"
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $23.60
    $23.60
    Log in for availability
    Case of 6
    Case of 6 - $134.70
    $134.70
    Each
       $23.60
    Case of 6
       $134.70
    Supplier Number: RS271/6