If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Size:6×50 µl
- Cat. no.:104016-406
DirectPlate XL™ DH10B Chemically Competent E.coli cells are a perfect choice for researchers looking to simplify their transformation workflow by eliminating heat shock, lengthy incubations, and time-consuming outgrowth procedures. Simply mix and directly plate, up to 250 kb.
- No heat shock
- Save time (3 minutes transformation instead of a 90 minute transformation)
- High transformation efficiency
- Clone 250 kb plus
Intact Genomics (ig®) DirectPlate-XL™ DH10B Competent cells offer simple, fast and robust results for your large DNA transformation needs – directly clone large DNA up to >250 kb. DirectPlate-XL™ DH10B chemically competent. DirectPlate-XL™ DH10B chemically competent E. coli cells provide higher transformation efficiency than any competitor's similar product and are suitable for high efficiency transformation in a wide variety of applications such as cloning and sub-cloning. Using proprietary technology, these cells can be used for transformation of extremely large DNA (i.e. >10 kb up to >250 kb).
The shipping and dry ice packaging cost is $45 for your entire order in the 48 contiguous United States.