Liquid Convection Apparatus, United Scientific
Catalog # 470332-614
Supplier: United Scientific Supplies
Specifications
- Description:Liquid convection apparatus
- Cat. no.:470332-614
Specifications
About this item
The liquid convection apparatus is designed to show the convection of heat in a liquid.
- 20 mm Ext.Ø. glass tube
- Tube is bent into 200×150 mm rectangle
The liquid convection apparatus is made from borosilicate glass, and is bent into a rectangle with a filling funnel. To show the convection of heat in a liquid, simply fill it with a colored liquid and place a heat source at one of the bottom corners. Glass tube is bent into a rectangle. Comes in a custom package to ensure product can be stored safely.
Required but not included are a support stand, clamp, heat source, and colored liquid.
Packaging: Supplied in custom package.