ContactUsLinkComponent
Coriolis Effect, United Scientific
Coriolis Effect, United Scientific
Catalog # 470332-612
undefined
Coriolis Effect, United Scientific
Catalog # 470332-612
Supplier Number:  UNCOEF001
Specifications
  • Description:
    Coriolis effect
  • Cat. no.:
    470332-612
Specifications
About this item
This apparatus helps students to understand the Coriolis effect, the phenomenon whereby an object moving on a spinning surface will travel in a curved path.
  • Components include:
  • Turntable
  • Erasable tracing surface
  • Ramp
  • Steel balls (2)
The demonstrations allow students to observe the Coriolis effect on a small scale, then explore how the Earth’s constant rotation influences wind patterns and ocean currents around the world.