Short Path Distillation Head, United Scientific Supplies
76531-658
- Description:Short path distillation head with coil condenser, 24/40 joints
- Environmentally Preferable:
- Cat. no.:76531-658
Short path Distillation Head with 24/40 flask joints and 10/30 thermometer port.
- Easy connect sockets 24/40
- Includes a port for a thermometer
Distillation head is manufactured from high quality 3.3 borosilicate glass and features a condenser coil and three ¼" hose nozzles, two for condenser and one for vacuum.