Pipettes, Transfer, Volumetric, Class A, Batch Certified, United Scientific Supplies
Supplier: United Scientific Supplies
Certificates
About this item
Volumetric pipettes with color-coding.
- Printed with durable amber markings
- Color-coded in blue for easy identification of capacity
Made from borosilicate glass in compliance with ASTM E969, Class A standards. Batch certified; calibrated to deliver.
Frequently Bought Together