Pipettes, Serological, Class A, Batch Certified, United Scientific Supplies
Supplier: United Scientific Supplies
Certificates
About this item
Type 3 serological pipettes with color-coding.
- Printed with durable amber markings at intervals
- Color-coded in white for easy identification of capacity
Made from borosilicate glass in compliance with USP, Class A standards. Batch certified and calibrated to deliver.
Customers Who Bought This Also Bought