Order Entry
Canada
Orders LinkContactUsLinkComponent
dNTP Set, Biotium

dNTP Set, Biotium

dNTP set, 100 mm each, 1 set - 250 µl each
Catalog # 10001-638
Supplier:   Biotium
Supplier #:  40052
dNTP Set, Biotium
Catalog # 10001-638
Supplier:  Biotium
CAS Number:  
dNTP Set, Biotium
dNTP Set, Biotium

dNTP set, 100 mm each, 1 set - 250 µl each

CAS Number:  
Price
$509.34
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Highly purified dNTPs (>99% by HPLC) in purified water. Each deoxynucleotide solution (dATP, dCTP, dGTP, dTTP) is provided in a separate vial at 100 mm.

The dNTPs are suitable for use in PCR, cDNA synthesis, sequencing, and other molecular biology applications.

Specifications

Specifications

  • Description:
    dNTP set, 100 mm each
  • Size:
    1 set - 250 µl each

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.