Abdos® Centrifuge Tubes with Conical Bottom, Sterile, Foxx Life Sciences
Supplier: Foxx Life Sciences
Certificates
About this item
These centrifuge tubes are manufactured using high purity virgin polypropylene (PP) conforming to US FDA 21 CFR free from natural rubber, heavy metal.
- Cap material: HDPE
- Silk-screen graduations
- 15000 RCF max.
- Color: Natural
- Temperature range: 0 to 135 °C
- Autoclavable: Yes
- Shelf life: 3 years
They are sterilized by gamma radiation.
Certifications: Conforms to US FDA 21 CFR. Complies with: ISO 11137. ISO Standard: 9001, 13485, 14001. Batch certificate included.
Packaging: Paper racked or bulk bag.
Frequently Bought Together