Specifications
- No. of Holes:80 (5×16)
- Color:Red
- Cat. no.:10011-292
- For Tubes:1.5/2.0 ml
- Supplier No.:R-80-R
Specifications
About this item
Microtube racks are available in three sizes and are designed to accommodate microtubes or screw-cap microtubes.
The 48-place aluminium rack holds 48 screw-cap tubes of all sizes. Unique ribbed-port design allows one-hand operation, supported by non-skid benchtop pads for a firm platform. Long-lasting aluminium construction. Centre support for added strength and stability.
The 80-well and 50-well racks hold eighty or fifty 1.5/2.0 ml micro and screw-cap microtubes, respectively. These racks resist most chemical and organic solvents and are perfect for storing tubes at cryogenic temperatures. Conical-bottom wells in 80-well racks allow superior stability of conical tubes. Each well is numbered for ease of sample identification. Frosted side writing patch. The 50-well racks feature smooth sides and are supplied within a clear plastic case.