ContactUsLinkComponent
Counting Chamber for Corning Cell Counter, 0.1 mm
Counting Chamber for Corning Cell Counter, 0.1 mm
Catalog # 76299-416
Supplier:  Corning
undefined
Counting Chamber for Corning Cell Counter, 0.1 mm
Catalog # 76299-416
Supplier:  Corning
Supplier Number:  480200
Specifications
  • Description:
    Counting chamber for Corning cell counter
  • Cat. no.:
    76299-416
  • Chamber Volume:
    10 µl
  • Chamber Depth:
    0.1 mm
Specifications
About this item
Counting Chamber for cell counter is a gridless slide with 0.1 mm depth same as a Neubauer Hemocytometer and comes standard with two high quality cover slip, notch for easy loading of cell suspension.