AxyPrep™ Mag plasmid purification Kit utilizes a magnetic bead technology for high throughput purification of plasmid DNA from E.coli cells. The AxyPrep™ Mag kit can also be used with fosmid and BAC vector-based constructs ranging from 5 to 150 Kb. The system consists of steps of pelleting cells, resuspension, lysis, neutralization, DNA binding, ethanol washing and plasmid elution. The protocol is automation-compatible.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Method / Format:Magnetic beads
- Prep size:Scalable
- Refine target molecule:Plasmid
- Sample type:Bacteria
- Target molecule:Plasmid
- Description:AxyPrep™ Mag plasmid purification kit