Order Entry
Canada
ContactUsLinkComponent
 
Influenza A H1N1 (A/swine/Cambridge/1939) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
 
Influenza A H1N1 (A/swine/Cambridge/1939) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    H1N1
  • Storage conditions:
    –20…–80 °C
  • Endotoxin content:
    <1.0 EU per μg protein as determined by the LAL method.
  • Reconstitution instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    Hemagglutinin
  • Protein/peptide name:
    HA
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    The recombinant hemagglutinin of the Influenza A virus (A/swine/Cambridge/1939 (H1N1)) consists of 518 amino acids and predicts a molecular mass of 58.5 kDa.
  • Sequence:
    Ala17-Pro523
  • Shipping temperature:
    Ambient
  • Gene ID:
    0

 

Frequently Bought Together