Human Recombinant respiratory syncytial virus (RSV) Fusion Protein (aa 1-526, His Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the Human respiratory syncytial virus (RSV) Fusion (AHX57537.1) (Met1-Met526) was expressed with a polyhistidine tag at the C-terminus.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:RSV
- Storage conditions:–20…–80 °C
- Endotoxin content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:AHX57537.1
- Reconstitution instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-free:Y
- Animal-free:Y
- Protein synonyms:fusion protein|HRSVgp08|F
- Protein/peptide name:RSV-F
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:The recombinant Human respiratory syncytial virus (RSV) Fusion consists of 512 amino acids and predicts a molecular mass of 57.04 kDa.
- Sequence:Met1-Met526
- Shipping temperature:Ambient
Specifications
Frequently Bought Together