Cynomolgus Recombinant FGL1 Protein (Fc Tag) (from HEK293 Cells)
A DNA sequence encoding the cynomolgus FGL1 (XP_005562748.1) (Met37-Ile348) was expressed with the Fc region of human IgG1 at the C-terminus.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Storage conditions:–20…–80 °C
- Endotoxin content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:XP_005562748.1
- Reconstitution instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-free:Y
- Animal-free:Y
- Protein synonyms:HP-041|HFREP1|LFIRE1|LFIRE-1
- Protein/peptide name:FGL1
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:The recombinant cynomolgus FGL1 comprises 495 amino acids and has a calculated molecular mass of 56.94 kDa.
- Sequence:Met37-Ile348
- Shipping temperature:Ambient
Frequently Bought Together