Cole-Parmer® Essentials Carboy with Handles, PP, FDA Compliant, Antylia Scientific
Supplier: Argos Technologies
Certificates
About this item
Get a better grip with two-handle construction.
- Leakproof polypropylene cap
- Easy to read, molded graduations
- 5000 ml graduation intervals
- Autoclavable at 250 °F (121 °C)
These rugged, durable plastic carboys will meet large volume liquid storage and dispensing needs. Integrated wide handles make them easy to carry and pour, even with gloves on. Round design maximizes mixing efficiency. Polypropylene cap with integrated seal ring ensures leakproof seal. Carboys are autoclavable at 250 °F (121 °C) and are chemical-resistant.
Customers Who Bought This Also Bought