Reusable Altessa Grid Pull-Over Cleanroom Hoods with Face Mask, Uniform Technology
Supplier: Uniform Technology
Certificates
About this item
Cleanroom hoods are manufactured from 99% polyester/1% carbon fabric.
- Pull-over hood with built-in face mask
- Vertical and horizontal snaps on back with elastic
Suitable for ISO Class 5 or higher cleanrooms.
Specifications
- Catalog No:
- 76515-942
- 76515-940
- 76515-938
- 76515-936
- 76515-934
- 76515-932
- 76515-930
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Fabric:
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon