VHB™ 3M™ LSE-110WF Tapes, TapeCase
Supplier: TAPECASE LTD. TE
Certificates
About this item
VHB™ Tapes are fast and easy-to-use, with a permanent bonding method to provide high strength and long-term durability.
- White, 45 mil (1.1 mm) thick, modified acrylic adhesive and conformable acrylic foam core
- Permanent bonding method provides high strength and long-term durability indoors and outdoors
- Maximum temperature: 302 °F (150 °C)
- Adhesive strength: 44 N/cm
VHB™ Tape LSE-110WF has modified acrylic adhesive on both sides of a conformable foam core. It can replace rivets, welds, and screws as well as other forms of mechanical fasteners or liquid adhesives.
Frequently Bought Together