VHB™ 3M™ 4955 Tapes, TapeCase
Supplier: TAPECASE LTD. TE
Certificates
About this item
VHB™ Tapes are fast and easy-to-use, with a permanent bonding method to provide high strength and long-term durability.
- White, 2.0 mm (80 mil), general-purpose adhesive and firm acrylic foam core
- Permanent bonding method provides high strength and long-term durability
- Can replace mechanical fasteners (rivets, welds, screws) or liquid adhesives
- Maximum temperature: 204 °C (400 °F)
- Adhesive strength: 20 lbs./inch
This white, 2.0 mm (0.080"), tape features a general-purpose acrylic adhesive with a firm foam core. It can replace rivets, welds, and screws. It also offers design flexibility with its viscoelasticity and powerful ability to bond to a variety of surfaces.
Customers Who Bought This Also Bought