Cole-Parmer® Essentials Graduated Rectangular Wide-Mouth HDPE Bottles, Antylia Scientific
Supplier: Argos Technologies
Certificates
About this item
Save valuable shelf and bench top space.
- Keep track of samples by attaching tag to closure ring
- Leakproof seal with included plastic insert
- Good chemical resistance
- Non-Autoclavable
Square and rectangular high-density polyethylene bottles provide good chemical resistance and heavy-duty service. Closure has small rings to attach tags for easy identification. Includes a plastic insert that ensures a leakproof seal.
BodyTextAccessories: Leakproof closure.
Customers Who Bought This Also Bought