ContactUsLinkComponent
United Scientific Supplies Beaker Set of 3, Borosilicate
United Scientific Supplies Beaker Set of 3, Borosilicate
  470332-658
undefined
United Scientific Supplies Beaker Set of 3, Borosilicate
  470332-658
 :  BGSET3
  • Capacity:
    250 / 400 / 600 ml
  • Description:
    Beakers, set of 3
  • Cat. no.:
    470332-658
An ideal starter set for any laboratory.
  • Double graduated metric scale and a marking area
  • Tolerance is approximately ±5%
This set contains three borosilicate glass beakers in sizes 250, 400, and 600 ml.